Assay Details
Target Gene Details
Entrez Gene ID: | 23172 |
Gene Name: | family with sequence similarity 175 member B |
Gene Aliases: |
ABRO1, KIAA0157 |
Location: |
Chr.10:124801785-124836670 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM175B | NM_032182.3 | NP_115558.3 | ||
| XM_011539554.1 | XP_011537856.1 | |||
| XM_017015986.1 | XP_016871475.1 | |||
| AK296677.1 | ||||
| AL537809.3 | ||||
| BC008999.1 | ||||
| D63877.1 | ||||
| DA148097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050837 | Chr.10:124678954 - 124828763 on Build GRCh38 | Gain |
|
| nsv832014 | Chr.10:124674236 - 124862247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map