Assay Details
Target Gene Details
Entrez Gene ID: | 51313 |
Gene Name: | family with sequence similarity 198 member B |
Gene Aliases: |
AD021, AD036, C4orf18 |
Location: |
Chr.4:158124580-158173050 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM198B | NM_001031700.2 | 6 | 4380 | NP_001026870.2 |
| NM_001128424.1 | 5 | 4356 | NP_001121896.1 | |
| NM_016613.6 | 5 | 4401 | NP_057697.2 | |
| AF260333.1 | 1 | 1842 | AAG44664.1 | |
| AK095474.1 | 1 | 1802 | ||
| AL832589.1 | 5 | 4227 | ||
| AL834177.1 | 5 | 3852 | CAD38874.1 | |
| BC017701.1 | 4 | 3304 | AAH17701.1 | |
| BC043193.2 | 6 | 4380 | AAH43193.1 | |
| BC100016.1 | 4 | 3240 | AAI00017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021067 | Chr.4:158051130 - 158254323 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145066523] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map