Assay Details
Target Gene Details
Entrez Gene ID: | 9118 |
Gene Name: | internexin neuronal intermediate filament protein alpha |
Gene Aliases: |
NEF5, NF-66, TXBP-1 |
Location: |
Chr.10:103277163-103290351 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INA | NM_032727.3 | 3 | 1792 | NP_116116.1 |
| AB209785.1 | 3 | 1243 | BAD93022.1 | |
| AK293490.1 | 3 | 806 | ||
| BC006359.2 | 3 | 1792 | AAH06359.1 | |
| S78296.1 | 3 | 1773 | AAB34482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509365 | Chr.10:103207187 - 103316390 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map