Assay Details
Target Gene Details
Entrez Gene ID: | 54443 |
Gene Name: | anillin actin binding protein |
Gene Aliases: |
FSGS8, Scraps, scra |
Location: |
Chr.7:36389803-36453791 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANLN | NM_001284301.2 | 2 | 312 | NP_001271230.1 |
| NM_001284302.2 | 2 | 312 | NP_001271231.1 | |
| NM_018685.4 | 2 | 312 | NP_061155.2 | |
| XM_006715746.1 | 2 | 309 | XP_006715809.1 | |
| XM_006715747.3 | 2 | 309 | XP_006715810.1 | |
| XM_017012354.1 | 2 | 309 | XP_016867843.1 | |
| XM_017012355.1 | 2 | 309 | XP_016867844.1 | |
| XM_017012356.1 | 2 | 309 | XP_016867845.1 | |
| AF273437.1 | 2 | 244 | AAF75796.1 | |
| AK291254.1 | 2 | 244 | ||
| AK299803.1 | 2 | 277 | ||
| BC070066.1 | 2 | 294 | AAH70066.1 | |
| BQ434804.1 | 2 | 292 |
More Information
Set Membership: |
|

Genomic Map