Assay Details
Target Gene Details
Entrez Gene ID: | 8048 |
Gene Name: | cysteine and glycine rich protein 3 |
Gene Aliases: |
CLP, CMD1M, CMH12, CRP3, LMO4, MLP |
Location: |
Chr.11:19182030-19210571 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRP3 | NM_003476.4 | 7 | 1133 | NP_003467.1 |
| AA192614.1 | 1 | 332 | ||
| AF121260.1 | AAF28868.1 | |||
| AI184002.1 | 1 | 331 | ||
| AJ711422.1 | 1 | 352 | ||
| BC005900.2 | 5 | 923 | AAH05900.1 | |
| BC024010.1 | AAH24010.1 | |||
| BC057221.1 | AAH57221.1 | |||
| U49837.1 | 6 | 951 | AAA92571.1 | |
| U72899.1 | AAD00183.1 |
More Information
Set Membership: |
|

Genomic Map