Assay Details
Target Gene Details
Entrez Gene ID: | 1997 |
Gene Name: | E74 like ETS transcription factor 1 |
Gene Aliases: |
- |
Location: |
Chr.13:40931919-41061414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF1 | NM_001145353.1 | NP_001138825.1 | ||
| NM_172373.3 | NP_758961.1 | |||
| XM_005266276.1 | XP_005266333.1 | |||
| XM_005266277.3 | XP_005266334.1 | |||
| XM_011534950.1 | XP_011533252.1 | |||
| XM_011534951.2 | XP_011533253.1 | |||
| XM_017020409.1 | XP_016875898.1 | |||
| XM_017020410.1 | XP_016875899.1 | |||
| XM_017020411.1 | XP_016875900.1 | |||
| XM_017020412.1 | XP_016875901.1 | |||
| XM_017020413.1 | XP_016875902.1 | |||
| AK304289.1 | ||||
| BC030507.2 | AAH30507.1 | |||
| BX640798.1 | CAE45881.1 | |||
| M82882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561538 | Chr.13:40940324 - 41070586 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map