Assay Details
Target Gene Details
Entrez Gene ID: | 6579 |
Gene Name: | solute carrier organic anion transporter family member 1A2 |
Gene Aliases: |
OATP, OATP-A, OATP1A2, SLC21A3 |
Location: |
Chr.12:21264600-21403657 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO1A2 | NM_021094.3 | 2 | 383 | NP_066580.1 |
| NM_134431.3 | 4 | 853 | NP_602307.1 | |
| XM_005253474.3 | 3 | 274 | XP_005253531.1 | |
| XM_005253477.3 | 2 | 161 | XP_005253534.1 | |
| XM_011520818.1 | 3 | 281 | XP_011519120.1 | |
| XM_011520819.1 | 3 | 428 | XP_011519121.1 | |
| XM_011520820.2 | 3 | 1750 | XP_011519122.1 | |
| XM_017019849.1 | 3 | 281 | XP_016875338.1 | |
| XM_017019850.1 | 2 | 161 | XP_016875339.1 | |
| AF085224.1 | 3 | 342 | AAD52694.1 | |
| AF279784.1 | 4 | 853 | AAG30037.1 | |
| AK296044.1 | ||||
| AK316249.1 | 3 | 546 | ||
| BC042452.1 | 2 | 163 | AAH42452.2 | |
| DA216595.1 | 3 | 285 | ||
| U21943.1 | 2 | 185 | AAA87732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557735 | Chr.12:21290348 - 21386023 on Build GRCh38 | Loss |
|
| esv3628799 | Chr.12:20916259 - 21333930 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map