Assay Details
Target Gene Details
Entrez Gene ID: | 6555 |
Gene Name: | solute carrier family 10 member 2 |
Gene Aliases: |
ASBT, IBAT, ISBT, NTCP2, PBAM |
Location: |
Chr.13:103043998-103066846 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A2 | NM_000452.2 | 6 | 1675 | NP_000443.1 |
| BC130521.1 | 6 | 1111 | ||
| BC130523.1 | 6 | 1111 | ||
| U10417.1 | 6 | 1676 | AAC51870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv363e214 | Chr.13:103031398 - 103052594 on Build GRCh38 | Gain |
|
| dgv3369n54 | Chr.13:103032716 - 103049023 on Build GRCh38 | Gain |
|
| dgv580e212 | Chr.13:103031333 - 103051790 on Build GRCh38 | Gain |
|
| nsv562981 | Chr.13:102820720 - 103931125 on Build GRCh38 | Gain |
|
| esv22017 | Chr.13:103031531 - 103049262 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map