Assay Details
Target Gene Details
Entrez Gene ID: | 153642 |
Gene Name: | arylsulfatase family member K |
Gene Aliases: |
TSULF |
Location: |
Chr.5:95555108-95605102 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSK | NM_198150.2 | 3 | 488 | NP_937793.1 |
| XM_005271904.3 | 3 | 501 | XP_005271961.1 | |
| AK303855.1 | 3 | 540 | ||
| AK316253.1 | 3 | 508 | ||
| AL832711.1 | 2 | 171 | CAD38634.1 | |
| AY358596.1 | 3 | 448 | AAQ88959.1 | |
| AY875939.1 | 3 | 330 | AAW66667.1 | |
| BC036047.1 | 3 | 350 | AAH36047.1 | |
| BC050392.1 | 3 | 447 | AAH50392.1 | |
| BC130329.1 | 3 | 426 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598955 | Chr.5:95567749 - 95800315 on Build GRCh38 | Gain |
|
| nsv830408 | Chr.5:95558328 - 95580291 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map