Assay Details
Target Gene Details
Entrez Gene ID: | 51574 |
Gene Name: | La ribonucleoprotein domain family member 7 |
Gene Aliases: |
ALAZS, HDCMA18P, PIP7S |
Location: |
Chr.4:112636964-112657592 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP7 | NM_001267039.1 | NP_001253968.1 | ||
| NM_015454.2 | NP_056269.1 | |||
| NM_016648.3 | NP_057732.2 | |||
| NR_049768.1 | ||||
| AF068284.1 | AAF65503.1 | |||
| AK000274.1 | ||||
| AK225900.1 | ||||
| AL049996.1 | CAB43230.1 | |||
| BC066945.1 | AAH66945.1 | |||
| BC107709.1 | ||||
| BX647100.1 | ||||
| EU667388.1 |
Target Gene Details
Entrez Gene ID: | 442912 |
Gene Name: | microRNA 367 |
Gene Aliases: |
MIRN367, hsa-mir-367 |
Location: |
Chr.4:112647874-112647941 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR367 | NR_029860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006030 | Chr.4:112618215 - 112736658 on Build GRCh38 | Gain |
|
| nsv525715 | Chr.4:112615590 - 112657453 on Build GRCh38 | Loss |
|
| nsv1014926 | Chr.4:112574087 - 112770975 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs578224265] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map