Assay Details
Target Gene Details
Entrez Gene ID: | 10236 |
Gene Name: | heterogeneous nuclear ribonucleoprotein R |
Gene Aliases: |
HNRPR, hnRNP-R |
Location: |
Chr.1:23304688-23344650 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPR | NM_001102397.2 | 7 | 899 | NP_001095867.1 |
| NM_001102398.2 | 8 | 1074 | NP_001095868.1 | |
| NM_001102399.2 | 7 | 908 | NP_001095869.1 | |
| NM_001297620.1 | 7 | 951 | NP_001284549.1 | |
| NM_001297621.1 | 6 | 722 | NP_001284550.1 | |
| NM_001297622.1 | 6 | 785 | NP_001284551.1 | |
| NM_005826.4 | 8 | 1065 | NP_005817.1 | |
| XM_005245711.4 | 8 | 987 | XP_005245768.1 | |
| XM_011540471.2 | 8 | 996 | XP_011538773.1 | |
| XM_011540472.2 | 6 | 746 | XP_011538774.1 | |
| XM_011540474.2 | 7 | 1249 | XP_011538776.1 | |
| XM_011540475.2 | 7 | 1246 | XP_011538777.1 | |
| XM_011540476.2 | 6 | 1080 | XP_011538778.1 | |
| XM_011540477.2 | 6 | 1017 | XP_011538779.1 | |
| XM_017000008.1 | 7 | 1240 | XP_016855497.1 | |
| XM_017000009.1 | 7 | 810 | XP_016855498.1 | |
| XM_017000010.1 | 7 | 825 | XP_016855499.1 | |
| AF000364.1 | 8 | 963 | AAC39540.1 | |
| AK297382.1 | 7 | 868 | ||
| AK297405.1 | 6 | 640 | ||
| AK300029.1 | 6 | 718 | ||
| AK311519.1 | 6 | 703 | ||
| AK313142.1 | 8 | 1013 | ||
| AU126696.1 | 3 | 354 | ||
| BC001449.2 | 8 | 1020 | AAH01449.1 | |
| BC110389.1 | 7 | 826 | ||
| BX640912.1 | 7 | 844 | CAE45953.1 | |
| BX647784.1 | 7 | 806 | ||
| DQ351905.1 | 6 | 788 | ABC73063.1 |
More Information
Set Membership: |
|

Genomic Map