Assay Details
Target Gene Details
Entrez Gene ID: | 1144 |
Gene Name: | cholinergic receptor nicotinic delta subunit |
Gene Aliases: |
ACHRD, CMS2A, CMS3A, CMS3B, CMS3C, FCCMS, SCCMS |
Location: |
Chr.2:232526160-232537907 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRND | NM_000751.2 | NP_000742.1 | ||
| NM_001256657.1 | NP_001243586.1 | |||
| NM_001311195.1 | NP_001298124.1 | |||
| NM_001311196.1 | NP_001298125.1 | |||
| XM_011510524.2 | XP_011508826.1 | |||
| AK291526.1 | ||||
| AK296704.1 | ||||
| AK300109.1 | ||||
| AK304890.1 | ||||
| AK315297.1 | ||||
| BC093923.1 | AAH93923.1 | |||
| BC093925.1 | AAH93925.1 | |||
| CD013895.1 | ||||
| X55019.1 | CAA38759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527789 | Chr.2:232436106 - 232533505 on Build GRCh38 | Loss |
|
| nsv821918 | Chr.2:232319063 - 232560965 on Build GRCh38 | Gain |
|
| nsv3212 | Chr.2:232526220 - 232564112 on Build GRCh38 | Deletion |
|
| nsv953202 | Chr.2:232474691 - 232546890 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map