Assay Details
Target Gene Details
Entrez Gene ID: | 1997 |
Gene Name: | E74 like ETS transcription factor 1 |
Gene Aliases: |
- |
Location: |
Chr.13:40931919-41061414 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF1 | NM_001145353.1 | 8 | 2200 | NP_001138825.1 |
| NM_172373.3 | 9 | 2417 | NP_758961.1 | |
| XM_005266276.1 | 9 | 2306 | XP_005266333.1 | |
| XM_005266277.3 | 9 | 2849 | XP_005266334.1 | |
| XM_011534950.1 | 11 | 2655 | XP_011533252.1 | |
| XM_011534951.2 | 10 | 2968 | XP_011533253.1 | |
| XM_017020409.1 | 11 | 2635 | XP_016875898.1 | |
| XM_017020410.1 | 10 | 2572 | XP_016875899.1 | |
| XM_017020411.1 | 10 | 2518 | XP_016875900.1 | |
| XM_017020412.1 | 10 | 2646 | XP_016875901.1 | |
| XM_017020413.1 | 10 | 2700 | XP_016875902.1 | |
| AK095466.1 | 1 | 729 | ||
| BC030507.2 | 8 | 2272 | AAH30507.1 | |
| BX640798.1 | 6 | 1698 | CAE45881.1 | |
| M82882.1 | 9 | 2359 |
More Information
Set Membership: |
|

Genomic Map