Assay Details
Target Gene Details
Entrez Gene ID: | 5465 |
Gene Name: | peroxisome proliferator activated receptor alpha |
Gene Aliases: |
NR1C1, PPAR, PPARalpha, hPPAR |
Location: |
Chr.22:46150534-46243756 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPARA | NM_001001928.2 | NP_001001928.1 | ||
| NM_005036.4 | NP_005027.2 | |||
| XM_005261655.3 | XP_005261712.1 | |||
| XM_006724269.3 | XP_006724332.1 | |||
| XM_006724270.3 | XP_006724333.1 | |||
| XM_011530239.2 | XP_011528541.1 | |||
| XM_011530240.2 | XP_011528542.1 | |||
| XM_011530241.2 | XP_011528543.1 | |||
| XM_011530244.2 | XP_011528546.1 | |||
| XM_011530245.2 | XP_011528547.1 | |||
| XM_017028839.1 | XP_016884328.1 | |||
| XM_017028840.1 | XP_016884329.1 | |||
| AF270490.1 | ||||
| AK289821.1 | ||||
| AU099251.1 | ||||
| AY258328.1 | AAO89523.1 | |||
| AY258330.1 | AAO89525.1 | |||
| L02932.1 | AAA36468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834224 | Chr.22:46111666 - 46261750 on Build GRCh38 | Loss |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map