Assay Details
Target Gene Details
Entrez Gene ID: | 3292 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 1 |
Gene Aliases: |
EDH17B2, EDHB17, HSD17, SDR28C1 |
Location: |
Chr.17:42551966-42555214 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B1 | NM_000413.2 | NP_000404.2 | ||
| XM_005257292.3 | XP_005257349.1 | |||
| XM_006721857.3 | XP_006721920.1 | |||
| XM_006721858.3 | XP_006721921.1 | |||
| XM_006721859.3 | XP_006721922.1 | |||
| XM_011524729.1 | XP_011523031.1 | |||
| XM_011524730.1 | XP_011523032.1 | |||
| XM_011524731.1 | XP_011523033.1 | |||
| XM_011524732.2 | XP_011523034.1 | |||
| AK127832.1 | ||||
| AK300143.1 | 2 | 452 | ||
| AK300159.1 | ||||
| AK300452.1 | ||||
| AK316292.1 | ||||
| BC033110.1 | 1 | 70 | AAH33110.1 | |
| BC104752.1 | AAI04753.1 | |||
| BC111935.1 | AAI11936.1 | |||
| DQ469954.1 | ||||
| M36263.1 | AAA35600.1 | |||
| X13440.1 | CAA31792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640608 | Chr.17:42549950 - 42556643 on Build GRCh38 | Loss |
|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
| esv30001 | Chr.17:42543879 - 42560687 on Build GRCh38 | Loss |
|
| nsv111876 | Chr.17:42549496 - 42555722 on Build GRCh38 | Deletion |
|
| nsv960106 | Chr.17:42549814 - 42556265 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map