Assay Details
Target Gene Details
Entrez Gene ID: | 5028 |
Gene Name: | purinergic receptor P2Y1 |
Gene Aliases: |
P2Y1 |
Location: |
Chr.3:152834693-152841439 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY1 | NM_002563.4 | 1 | 2176 | NP_002554.1 |
| AF018284.1 | 1 | 804 | AAB94556.1 | |
| AK313920.1 | ||||
| AY136752.1 | AAN01278.1 | |||
| BC074784.2 | 1 | 1108 | AAH74784.1 | |
| BC074785.2 | 1 | 1108 | AAH74785.1 | |
| U42029.1 | 1 | 1132 | AAA97872.1 | |
| U42030.1 | 1 | 1223 | AAA97873.1 | |
| Z49205.1 | 1 | 1936 | CAA89066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998184 | Chr.3:152675973 - 153414946 on Build GRCh38 | Gain |
|
| nsv1008013 | Chr.3:152644272 - 152889874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map