Assay Details
Target Gene Details
Entrez Gene ID: | 8562 |
Gene Name: | density regulated re-initiation and release factor |
Gene Aliases: |
DRP, DRP1, SMAP-3 |
Location: |
Chr.12:122752821-122771064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENR | NM_003677.4 | NP_003668.2 | ||
| AB014731.1 | BAB20268.1 | |||
| AF038554.1 | AAC02985.2 | |||
| AF103800.1 | AAF02420.1 | |||
| AK299463.1 | ||||
| BC007860.2 | AAH07860.1 | |||
| BP872988.1 |
More Information
Set Membership: |
|

Genomic Map