Assay Details
Target Gene Details
Entrez Gene ID: | 7570 |
Gene Name: | zinc finger protein 22 |
Gene Aliases: |
HKR-T1, KOX15, ZNF422, Zfp422 |
Location: |
Chr.10:45000825-45005329 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF22 | NM_006963.4 | 2 | 511 | NP_008894.2 |
| AK291508.1 | 2 | 413 | ||
| AK312278.1 | 2 | 413 | ||
| BC010642.1 | 2 | 401 | AAH10642.1 | |
| BC041139.1 | 2 | 862 | AAH41139.1 | |
| BC053687.1 | 2 | 405 | AAH53687.1 | |
| BT009806.1 | 1 | 268 | AAP88808.1 | |
| S50223.1 | 1 | 357 | AAB24264.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576560 | Chr.10:44916005 - 45054697 on Build GRCh38 | Gain |
|
| esv3623079 | Chr.10:44993759 - 45028111 on Build GRCh38 | Loss |
|
| nsv831848 | Chr.10:44994728 - 45172222 on Build GRCh38 | Gain |
|
| nsv831846 | Chr.10:44826788 - 45060922 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map