Assay Details
Target Gene Details
Entrez Gene ID: | 1844 |
Gene Name: | dual specificity phosphatase 2 |
Gene Aliases: |
PAC-1, PAC1 |
Location: |
Chr.2:96143166-96145468 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP2 | NM_004418.3 | NP_004409.1 | ||
| XM_017003546.1 | XP_016859035.1 | |||
| AF331843.1 | 1 | 450 | AAL57044.1 | |
| BC007771.2 | AAH07771.1 | |||
| L11329.1 | AAA50779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
|
| dgv4009n100 | Chr.2:96072837 - 96159625 on Build GRCh38 | Gain |
|
| nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
|
| esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map