Assay Details
Target Gene Details
Entrez Gene ID: | 51720 |
Gene Name: | ubiquitin interaction motif containing 1 |
Gene Aliases: |
RAP80, X2HRIP110 |
Location: |
Chr.5:176905005-177022640 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UIMC1 | NM_001199297.2 | 16 | 2583 | NP_001186226.1 |
| NM_001199298.1 | 15 | 2358 | NP_001186227.1 | |
| NM_001317961.1 | 15 | 1971 | NP_001304890.1 | |
| NM_016290.4 | 15 | 2372 | NP_057374.3 | |
| XM_005265930.2 | 15 | 2346 | XP_005265987.1 | |
| XM_005265932.2 | 13 | 1482 | XP_005265989.1 | |
| XM_005265933.2 | 13 | 1594 | XP_005265990.1 | |
| XM_005265934.2 | 15 | 1801 | XP_005265991.1 | |
| XM_005265935.2 | 15 | 1810 | XP_005265992.1 | |
| XM_005265936.2 | 14 | 1600 | XP_005265993.1 | |
| XM_006714871.2 | 15 | 2321 | XP_006714934.1 | |
| XM_006714874.2 | 15 | 1785 | XP_006714937.1 | |
| XM_017009574.1 | 14 | 2237 | XP_016865063.1 | |
| XM_017009575.1 | 16 | 2049 | XP_016865064.1 | |
| XM_017009576.1 | 13 | 1495 | XP_016865065.1 | |
| XM_017009577.1 | 13 | 1547 | XP_016865066.1 | |
| XM_017009578.1 | 14 | 1609 | XP_016865067.1 | |
| XM_017009579.1 | 14 | 1509 | XP_016865068.1 | |
| AF113538.1 | 11 | 1162 | AAF14875.1 | |
| AF349313.1 | 15 | 2314 | AAK61871.1 | |
| AK023044.1 | 16 | 2360 | ||
| AL137375.1 | 6 | 690 | CAB70716.1 | |
| BC006078.1 | 15 | 2279 | AAH06078.1 | |
| BC032561.1 | 15 | 1762 | AAH32561.1 | |
| BX537376.1 | 15 | 2372 | CAD97618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016283 | Chr.5:176904803 - 176968410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map