Assay Details
Target Gene Details
Entrez Gene ID: | 8899 |
Gene Name: | pre-mRNA processing factor 4B |
Gene Aliases: |
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1 |
Location: |
Chr.6:4019402-4064983 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
More Information
Set Membership: |
|

Genomic Map