Assay Details
Target Gene Details
Entrez Gene ID: | 59338 |
Gene Name: | pleckstrin homology domain containing A1 |
Gene Aliases: |
TAPP1 |
Location: |
Chr.10:122374578-122432355 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042640 | Chr.10:122369174 - 122442149 on Build GRCh38 | Loss |
|
| dgv987n100 | Chr.10:122379070 - 122437149 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map