Assay Details
Target Gene Details
Entrez Gene ID: | 5771 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 2 |
Gene Aliases: |
PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP |
Location: |
Chr.18:12785478-12884351 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN2 | NM_001207013.1 | NP_001193942.1 | ||
| NM_001308287.1 | 8 | 1630 | NP_001295216.1 | |
| NM_002828.3 | 9 | 1735 | NP_002819.2 | |
| NM_080422.2 | NP_536347.1 | |||
| NM_080423.2 | NP_536348.1 | |||
| XM_005258124.3 | 10 | 1802 | XP_005258181.1 | |
| XM_005258125.3 | XP_005258182.1 | |||
| XM_011525705.2 | 11 | 2022 | XP_011524007.1 | |
| XM_011525706.2 | 8 | 1523 | XP_011524008.1 | |
| XM_011525707.2 | 5 | 1130 | XP_011524009.1 | |
| XM_017025884.1 | XP_016881373.1 | |||
| XM_017025885.1 | 10 | 1937 | XP_016881374.1 | |
| XM_017025886.1 | 8 | 1625 | XP_016881375.1 | |
| XM_017025887.1 | XP_016881376.1 | |||
| XM_017025888.1 | XP_016881377.1 | |||
| AB209569.1 | 8 | 1662 | BAD92806.1 | |
| AK225706.1 | ||||
| AK290649.1 | ||||
| BC008244.1 | AAH08244.1 | |||
| BC016727.1 | AAH16727.1 | |||
| M25393.1 | 9 | 1601 | AAA65997.1 | |
| M80737.1 | ||||
| M81478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064273 | Chr.18:12697602 - 12828252 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map