Assay Details
Target Gene Details
Entrez Gene ID: | 51213 |
Gene Name: | leucine zipper protein 4 |
Gene Aliases: |
CT-28, CT-8, CT28, HOM-TES-85 |
Location: |
Chr.X:115289717-115307556 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP4 | NM_001318840.1 | 3 | 1518 | NP_001305769.1 |
| NM_016383.4 | 4 | 1637 | NP_057467.1 | |
| AF124430.1 | 4 | 1607 | AAF28870.1 | |
| AK093369.1 | 3 | 1509 | ||
| AW118228.1 | 1 | 110 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577051 | Chr.X:115223253 - 115839825 on Build GRCh38 | Gain |
|
| nsv520351 | Chr.X:115073935 - 115318424 on Build GRCh38 | Gain |
|
| esv33994 | Chr.X:113685854 - 115738426 on Build GRCh38 | Gain+Loss |
|
| dgv65n50 | Chr.X:115306992 - 115310755 on Build GRCh38 | Loss |
|
| esv2666748 | Chr.X:115307080 - 115310127 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map