Assay Details
Target Gene Details
Entrez Gene ID: | 3421 |
Gene Name: | isocitrate dehydrogenase 3 (NAD(+)) gamma |
Gene Aliases: |
H-IDHG |
Location: |
Chr.X:153785766-153794523 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH3G | NM_004135.3 | NP_004126.1 | ||
| NM_174869.2 | 12 | 1305 | NP_777358.1 | |
| AB209206.1 | 6 | 3782 | BAD92443.1 | |
| BC000933.2 | AAH00933.1 | |||
| BC001902.1 | AAH01902.1 | |||
| U40272.1 | AAD09357.1 | |||
| U69268.1 | 8 | 856 | AAB70115.1 | |
| U69269.1 | AAB70116.1 | |||
| Z68907.1 | CAA93143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map