Assay Details
Target Gene Details
Entrez Gene ID: | 8688 |
Gene Name: | keratin 37 |
Gene Aliases: |
HA7, K37, KRTHA7 |
Location: |
Chr.17:41420557-41424570 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT37 | NM_003770.4 | NP_003761.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map