Assay Details
Target Gene Details
Entrez Gene ID: | 440145 |
Gene Name: | mitotic spindle organizing protein 1 |
Gene Aliases: |
C13orf37, MOZART1 |
Location: |
Chr.13:72708357-72727800 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MZT1 | NM_001071775.2 | NP_001065243.1 | ||
| AK025522.1 | ||||
| BC125183.1 |
More Information
Set Membership: |
|

Genomic Map