Assay Details
Target Gene Details
Entrez Gene ID: | 202 |
Gene Name: | absent in melanoma 1 |
Gene Aliases: |
CRYBG1, ST4 |
Location: |
Chr.6:106360808-106570449 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM1 | NM_001624.3 | NP_001615.2 | ||
| XM_005266839.3 | XP_005266896.1 | |||
| XM_017010333.1 | XP_016865822.1 | |||
| AK025693.1 | ||||
| AK297008.1 | ||||
| AK300443.1 | ||||
| AK316287.1 | ||||
| U83115.1 | AAB53791.1 |
Target Gene Details
Entrez Gene ID: | 84816 |
Gene Name: | reticulon 4 interacting protein 1 |
Gene Aliases: |
NIMP, OPA10 |
Location: |
Chr.6:106559237-106630921 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN4IP1 |
More Information
Set Membership: |
|

Genomic Map