Assay Details
Target Gene Details
Entrez Gene ID: | 200894 |
Gene Name: | ADP ribosylation factor like GTPase 13B |
Gene Aliases: |
ARL2L1, JBTS8 |
Location: |
Chr.3:93980139-94055678 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL13B | NM_001174150.1 | NP_001167621.1 | ||
| NM_001174151.1 | NP_001167622.1 | |||
| NM_001321328.1 | NP_001308257.1 | |||
| NM_144996.3 | NP_659433.2 | |||
| NM_182896.2 | NP_878899.1 | |||
| NR_033427.1 | ||||
| NR_135621.1 | ||||
| XM_006713532.3 | XP_006713595.1 | |||
| XM_011512532.2 | XP_011510834.1 | |||
| XM_011512533.2 | XP_011510835.1 | |||
| XM_011512534.2 | XP_011510836.1 | |||
| XM_011512535.2 | XP_011510837.1 | |||
| XM_017005853.1 | XP_016861342.1 | |||
| AK296994.1 | ||||
| AK299284.1 | ||||
| AL713789.1 | CAD28544.2 | |||
| BC094725.1 | AAH94725.1 | |||
| BC104035.1 | AAI04036.1 | |||
| BC104036.1 | AAI04037.1 | |||
| BG705058.1 | ||||
| CR936753.1 | ||||
| CR936756.1 | ||||
| CR936763.1 |
Target Gene Details
Entrez Gene ID: | 415117 |
Gene Name: | syntaxin 19 |
Gene Aliases: |
- |
Location: |
Chr.3:94014365-94029281 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX19 | NM_001001850.2 | 2 | 676 | NP_001001850.1 |
| XM_011512842.2 | 3 | 1407 | XP_011511144.1 | |
| AF461456.1 | 1 | 419 | AAQ04780.1 | |
| BC034696.1 | 2 | 676 | AAH34696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4814n100 | Chr.3:93800622 - 94025077 on Build GRCh38 | Gain |
|
| nsv590987 | Chr.3:93989976 - 94060820 on Build GRCh38 | Loss |
|
| esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map