Assay Details
Target Gene Details
Entrez Gene ID: | 171484 |
Gene Name: | family with sequence similarity 9 member C |
Gene Aliases: |
TEX39C |
Location: |
Chr.X:13035617-13044798 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM9C | NM_174901.5 | NP_777561.1 | ||
| XM_005274460.2 | XP_005274517.1 | |||
| XM_017029321.1 | XP_016884810.1 | |||
| AF494345.1 | AAN07164.1 | |||
| AK313776.1 | ||||
| BX647580.1 | CAI46094.1 | |||
| BX648111.1 | CAI46034.1 | |||
| BX648919.1 | CAI46007.1 | |||
| DA958431.1 |
More Information
Set Membership: |
|

Genomic Map