Assay Details
Target Gene Details
Entrez Gene ID: | 27101 |
Gene Name: | calcyclin binding protein |
Gene Aliases: |
GIG5, PNAS-107, S100A6BP, SIP |
Location: |
Chr.1:174999435-175012027 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACYBP | NM_001007214.1 | 6 | 1749 | NP_001007215.1 |
| NM_014412.2 | 6 | 1934 | NP_055227.1 | |
| AK093425.1 | 6 | 1721 | ||
| AL035305.1 | 6 | 1626 | CAA22910.1 |
More Information
Set Membership: |
|

Genomic Map