Assay Details
Target Gene Details
Entrez Gene ID: | 57477 |
Gene Name: | shroom family member 4 |
Gene Aliases: |
SHAP |
Location: |
Chr.X:50575534-50814167 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHROOM4 | NM_020717.3 | 9 | 9055 | NP_065768.2 |
| NR_027121.1 | ||||
| AB033028.1 | ||||
| BC151240.1 |
More Information
Set Membership: |
|

Genomic Map