Assay Details
Target Gene Details
Entrez Gene ID: | 3755 |
Gene Name: | potassium voltage-gated channel modifier subfamily G member 1 |
Gene Aliases: |
K13, KCNG, KV6.1, kH2 |
Location: |
Chr.20:51003656-51023176 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNG1 | NM_002237.3 | 3 | 1726 | NP_002228.2 |
| XM_006723785.2 | 5 | 2109 | XP_006723848.1 | |
| XM_011528800.1 | 4 | 1784 | XP_011527102.1 | |
| XM_011528801.1 | 5 | 1893 | XP_011527103.1 | |
| XM_011528802.1 | 5 | 1888 | XP_011527104.1 | |
| XM_011528803.2 | 5 | 1815 | XP_011527105.1 | |
| XM_011528804.1 | 4 | 2038 | XP_011527106.1 | |
| XM_011528805.2 | 3 | 3949 | XP_011527107.1 | |
| XM_011528806.1 | 3 | 1930 | XP_011527108.1 | |
| AF033383.1 | 2 | 1431 | AAC05635.1 | |
| AK290689.1 | 3 | 1727 | ||
| AW162513.1 | 1 | 501 | ||
| BC046629.2 | 3 | 1695 | AAH46629.1 | |
| BG029104.1 | 1 | 595 | ||
| CN480356.1 |
More Information
Set Membership: |
|

Genomic Map