Assay Details
Target Gene Details
Entrez Gene ID: | 51744 |
Gene Name: | CD244 molecule |
Gene Aliases: |
2B4, NAIL, NKR2B4, Nmrk, SLAMF4 |
Location: |
Chr.1:160830160-160862902 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD244 | NM_001166663.1 | 9 | 2420 | NP_001160135.1 |
| NM_001166664.1 | 8 | 2129 | NP_001160136.1 | |
| NM_016382.3 | 9 | 2405 | NP_057466.1 | |
| XM_011509623.2 | 7 | 1805 | XP_011507925.1 | |
| AF117711.1 | 9 | 2359 | AAF28833.1 | |
| AF145782.1 | 9 | 2180 | AAD38951.1 | |
| BC028073.1 | AAH28073.1 | |||
| BC053985.1 | 9 | 2191 | AAH53985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548057 | Chr.1:160777825 - 160882256 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78916399] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map