Assay Details
Target Gene Details
Entrez Gene ID: | 221120 |
Gene Name: | alkB homolog 3, alpha-ketoglutaratedependent dioxygenase |
Gene Aliases: |
ABH3, DEPC-1, DEPC1, PCA1, hABH3 |
Location: |
Chr.11:43880807-43920275 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH3 | NM_139178.3 | 10 | 1243 | NP_631917.1 |
| AB042029.1 | 10 | 1206 | BAB70508.1 | |
| AK125602.1 | 1 | 2302 | ||
| AK315266.1 | 10 | 1310 | ||
| BC015155.1 | 5 | 529 | AAH15155.1 | |
| BC067257.1 | 10 | 1240 | AAH67257.1 | |
| BC103812.1 | 10 | 981 | AAI03813.1 | |
| BC103813.1 | 10 | 981 | AAI03814.1 | |
| BC103814.2 | 10 | 981 | AAI03815.1 | |
| CD364334.1 | 1 | 343 |
Target Gene Details
Entrez Gene ID: | 100507300 |
Gene Name: | ALKBH3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:43909289-43920944 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH3-AS1 | NR_038907.1 | |||
| AK097541.1 | ||||
| BI827816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv554171 | Chr.11:43897861 - 43927961 on Build GRCh38 | Gain |
|
| nsv832138 | Chr.11:43778980 - 43966454 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map