Assay Details
Target Gene Details
Entrez Gene ID: | 84676 |
Gene Name: | tripartite motif containing 63 |
Gene Aliases: |
IRF, MURF1, MURF2, RNF28, SMRZ |
Location: |
Chr.1:26051304-26067634 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM63 | NM_032588.3 | 9 | 1396 | NP_115977.2 |
| AF353673.1 | 9 | 1375 | AAK39519.1 | |
| AF361946.1 | 4 | 1537 | AAK52497.1 | |
| AJ276484.1 | 9 | 1379 | CAC81706.1 | |
| AK056942.1 | 9 | 1392 | BAB71318.1 | |
| AK297820.1 | ||||
| BC080529.1 | 9 | 1374 | AAH80529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834513 | Chr.1:25922980 - 26087385 on Build GRCh38 | Gain |
|
| nsv508678 | Chr.1:26051066 - 26106136 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map