Assay Details
Target Gene Details
Entrez Gene ID: | 6878 |
Gene Name: | TATA-box binding protein associated factor 6 |
Gene Aliases: |
MGC:8964, TAF(II)70, TAF(II)80, TAF2E, TAFII-70, TAFII-80, TAFII70, TAFII80, TAFII85 |
Location: |
Chr.7:100107070-100119858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF6 | NM_001190415.1 | NP_001177344.1 | ||
| NM_005641.3 | NP_005632.1 | |||
| NM_139315.2 | NP_647476.1 | |||
| NR_033792.1 | ||||
| XM_006716100.2 | XP_006716163.1 | |||
| XM_006716101.2 | XP_006716164.1 | |||
| XM_006716102.2 | XP_006716165.1 | |||
| XM_011516542.2 | XP_011514844.1 | |||
| XM_011516543.2 | XP_011514845.1 | |||
| XM_017012568.1 | XP_016868057.1 | |||
| AK097781.1 | ||||
| AK300005.1 | ||||
| AK314428.1 | ||||
| AK315077.1 | ||||
| BC018115.1 | AAH18115.1 | |||
| CR627390.1 | CAH10485.1 | |||
| L25444.1 | AAA63643.1 | |||
| U31659.1 | AAA84390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| nsv951384 | Chr.7:100100678 - 100114877 on Build GRCh38 | Deletion |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map