Assay Details
Target Gene Details
Entrez Gene ID: | 669 |
Gene Name: | bisphosphoglycerate mutase |
Gene Aliases: |
DPGM |
Location: |
Chr.7:134646779-134679816 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPGM | NM_001293085.1 | 4 | 1661 | NP_001280014.1 |
| NM_001724.4 | 3 | 1536 | NP_001715.1 | |
| NM_199186.2 | 4 | 1857 | NP_954655.1 | |
| XM_011516527.1 | 3 | 1717 | XP_011514829.1 | |
| AK097778.1 | 3 | 1460 | ||
| BC017050.1 | 3 | 1507 | AAH17050.1 | |
| BU608668.1 | 1 | 265 | ||
| X04327.1 | 3 | 1426 | CAA27858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762705 | Chr.7:134667005 - 134694885 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs184837736] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map