Assay Details
Target Gene Details
Entrez Gene ID: | 8687 |
Gene Name: | keratin 38 |
Gene Aliases: |
HA8, KRTHA8, hHa8 |
Location: |
Chr.17:41436369-41441344 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT38 | NM_006771.3 | NP_006762.3 | ||
| AJ786656.1 | CAH10351.1 | |||
| BC131715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map