Assay Details
Target Gene Details
Entrez Gene ID: | 90806 |
Gene Name: | angel homolog 2 |
Gene Aliases: |
Ccr4d, KIAA0759L |
Location: |
Chr.1:212992182-213015875 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL2 | NM_001300753.1 | 3 | 628 | NP_001287682.1 |
| NM_001300755.1 | 3 | 726 | NP_001287684.1 | |
| NM_001300757.1 | 2 | 381 | NP_001287686.1 | |
| NM_001300758.1 | 2 | 377 | NP_001287687.1 | |
| NM_144567.4 | 3 | 703 | NP_653168.2 | |
| NR_125333.1 | 2 | 377 | ||
| XM_005273344.1 | 3 | 724 | XP_005273401.1 | |
| XM_005273345.1 | 3 | 770 | XP_005273402.1 | |
| XM_005273346.2 | 3 | 523 | XP_005273403.1 | |
| XM_005273347.3 | 2 | 395 | XP_005273404.1 | |
| XM_017002774.1 | 3 | 652 | XP_016858263.1 | |
| XM_017002775.1 | 4 | 951 | XP_016858264.1 | |
| XM_017002776.1 | 4 | 975 | XP_016858265.1 | |
| XM_017002777.1 | 4 | 1021 | XP_016858266.1 | |
| XM_017002778.1 | 4 | 764 | XP_016858267.1 | |
| AF510741.1 | 2 | 332 | AAM44053.1 | |
| AK294479.1 | 3 | 502 | ||
| AK295116.1 | 3 | 632 | ||
| AK299208.1 | 3 | 695 | ||
| AK299365.1 | 2 | 287 | ||
| AK311276.1 | 2 | 286 | ||
| AL079275.1 | 3 | 658 | ||
| AY468376.1 | 2 | 433 | ||
| BC016966.1 | 2 | 447 | AAH16966.1 | |
| BC047469.1 | 2 | 293 | AAH47469.1 |
More Information
Set Membership: |
|

Genomic Map