Assay Details
Target Gene Details
Entrez Gene ID: | 9455 |
Gene Name: | homer scaffolding protein 2 |
Gene Aliases: |
ACPD, CPD, DFNA68, HOMER-2, VESL-2 |
Location: |
Chr.15:82836946-82986176 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOMER2 | NM_004839.3 | 9 | 1687 | NP_004830.2 |
| NM_199330.2 | 9 | 1720 | NP_955362.1 | |
| XM_005272448.4 | 9 | 2155 | XP_005272505.1 | |
| XM_005272449.3 | 9 | 1567 | XP_005272506.1 | |
| XM_006720775.3 | XP_006720838.1 | |||
| XM_006720776.3 | 9 | 1623 | XP_006720839.1 | |
| XM_011522231.2 | 9 | 2039 | XP_011520533.1 | |
| XM_011522232.2 | XP_011520534.1 | |||
| XM_011522233.2 | 9 | 1570 | XP_011520535.1 | |
| XM_011522234.2 | 9 | 1837 | XP_011520536.1 | |
| AF093263.1 | 8 | 1498 | AAC71027.1 | |
| AF093264.1 | 8 | 1531 | AAC71028.1 | |
| BC012109.1 | 9 | 1684 | AAH12109.1 | |
| CB321822.1 | 1 | 290 | ||
| CR936744.1 | ||||
| Y19025.1 | 8 | 1498 | CAB75537.1 | |
| Y19026.1 | 8 | 1531 | CAB75538.1 | |
| Y19027.1 | 8 | 1334 | CAB75539.1 | |
| Y19028.1 | 8 | 1367 | CAB75540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469557 | Chr.15:82749011 - 82891877 on Build GRCh38 | Loss |
|
| nsv471687 | Chr.15:82749012 - 82891877 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map