Assay Details
Target Gene Details
Entrez Gene ID: | 90288 |
Gene Name: | EF-hand calcium binding domain 12 |
Gene Aliases: |
C3orf25 |
Location: |
Chr.3:129401321-129428651 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB12 | NM_207307.2 | 4 | 916 | NP_997190.1 |
| XM_011513293.2 | 4 | 2990 | XP_011511595.1 | |
| XM_011513294.2 | 4 | 2988 | XP_011511596.1 | |
| XM_011513295.2 | 4 | 2991 | XP_011511597.1 | |
| XM_011513297.2 | 4 | 2992 | XP_011511599.1 | |
| AK096099.1 | 2 | 342 | ||
| AK127968.1 | 4 | 901 | ||
| AL133011.1 | 3 | 484 | CAH10712.1 | |
| BC066975.1 | 4 | 916 | AAH66975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2725901 | Chr.3:129380821 - 129487939 on Build GRCh38 | Deletion |
|
| esv3597741 | Chr.3:129383049 - 129486193 on Build GRCh38 | Loss |
|
| nsv591668 | Chr.3:129403449 - 129490335 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map