Assay Details
Target Gene Details
Entrez Gene ID: | 9941 |
Gene Name: | endo/exonuclease (5'-3'), endonuclease G-like |
Gene Aliases: |
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB |
Location: |
Chr.3:38496127-38526305 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Exon 10 |
More Information
Set Membership: |
|

Genomic Map