Assay Details
Target Gene Details
Entrez Gene ID: | 55081 |
Gene Name: | intraflagellar transport 57 |
Gene Aliases: |
ESRRBL1, HIPPI, MHS4R2 |
Location: |
Chr.3:108160812-108222570 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT57 | NM_018010.3 | 11 | 1472 | NP_060480.1 |
| AB014762.1 | 11 | 1224 | BAB87803.1 | |
| AF139576.1 | 11 | 1302 | AAK13588.1 | |
| AF245220.1 | 11 | 1366 | AAK28373.1 | |
| AK001009.1 | 11 | 1311 | BAA91466.1 | |
| AK298854.1 | 11 | 1370 | ||
| BC009690.2 | 7 | 639 | AAH09690.2 | |
| BC011899.2 | 11 | 1321 | AAH11899.1 | |
| CR457251.1 | 11 | 1224 | CAG33532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011158 | Chr.3:107746970 - 108629120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map