Assay Details
Target Gene Details
Entrez Gene ID: | 161253 |
Gene Name: | RRAD and GEM like GTPase 2 |
Gene Aliases: |
- |
Location: |
Chr.14:22883165-22887680 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REM2 | NM_173527.2 | 5 | 1576 | NP_775798.2 |
| XM_005267383.3 | 5 | 1637 | XP_005267440.1 | |
| XM_017021058.1 | 4 | 2786 | XP_016876547.1 | |
| XM_017021059.1 | 4 | 2783 | XP_016876548.1 | |
| AK096283.1 | 5 | 1576 | BAC04746.1 | |
| BC035663.1 | 5 | 1567 | AAH35663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1214 | Chr.14:22877852 - 22890492 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map