Assay Details
Target Gene Details
Entrez Gene ID: | 7565 |
Gene Name: | zinc finger protein 17 |
Gene Aliases: |
HPF3, KOX10 |
Location: |
Chr.19:57409252-57421939 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF17 | NM_006959.2 | 3 | 2244 | NP_008890.2 |
| XM_017027205.1 | 4 | 4194 | XP_016882694.1 | |
| AB075827.1 | 1 | 1851 | BAB85533.1 | |
| AK097123.1 | 1 | 1530 | BAC04956.1 | |
| AK098183.1 | 3 | 2226 | BAC05252.1 | |
| AK127948.1 | 4 | 2283 | ||
| AK128471.1 | 1 | 2449 |
More Information
Set Membership: |
|

Genomic Map