Assay Details
Target Gene Details
Entrez Gene ID: | 148789 |
Gene Name: | beta-1,3-N-acetylgalactosaminyltransferase 2 |
Gene Aliases: |
B3GalNAc-T2, MDDGA11 |
Location: |
Chr.1:235440654-235504481 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALNT2 | NM_152490.4 | 12 | 1786 | NP_689703.1 |
| XM_006711749.3 | XP_006711812.1 | |||
| XM_017000394.1 | 13 | 1909 | XP_016855883.1 | |
| XM_017000395.1 | 13 | 1873 | XP_016855884.1 | |
| AB209046.1 | 11 | 1340 | BAD92283.1 | |
| BC029564.1 | 12 | 1786 | AAH29564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013571 | Chr.1:235210731 - 235587805 on Build GRCh38 | Gain |
|
| dgv30n21 | Chr.1:235198002 - 235605078 on Build GRCh38 | Gain |
|
| esv3575794 | Chr.1:235370375 - 235549222 on Build GRCh38 | Gain |
|
| nsv549392 | Chr.1:235375705 - 235548388 on Build GRCh38 | Gain |
|
| nsv1011288 | Chr.1:235376656 - 235666302 on Build GRCh38 | Gain |
|
| nsv468406 | Chr.1:235395391 - 235463092 on Build GRCh38 | Gain |
|
| dgv586n100 | Chr.1:235367709 - 235547476 on Build GRCh38 | Gain |
|
| dgv587n100 | Chr.1:235445295 - 235531886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map