Assay Details
Target Gene Details
Entrez Gene ID: | 57113 |
Gene Name: | transient receptor potential cation channel subfamily C member 7 |
Gene Aliases: |
TRP7 |
Location: |
Chr.5:136213311-136365476 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC7 | NM_001167576.1 | NP_001161048.1 | ||
| NM_001167577.1 | NP_001161049.1 | |||
| NM_020389.2 | NP_065122.1 | |||
| AJ272034.1 | CAC03489.1 | |||
| AJ421783.1 | CAD19069.1 | |||
| AJ549088.1 | CAD70161.1 | |||
| AJ549089.1 | CAD70162.1 | |||
| AJ549090.1 | CAD70163.1 | |||
| AY167926.1 | AAO12124.1 | |||
| AY167927.1 | AAO12125.1 | |||
| BC128185.1 | ||||
| BC128186.1 | ||||
| BI826927.1 |
More Information
Set Membership: |
|

Genomic Map