Assay Details
Target Gene Details
Entrez Gene ID: | 8099 |
Gene Name: | cyclin dependent kinase 2 associated protein 1 |
Gene Aliases: |
DOC1, DORC1, ST19, doc-1, p12DOC-1 |
Location: |
Chr.12:123260970-123272316 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK2AP1 | NM_001270433.1 | 4 | 934 | NP_001257362.1 |
| NM_001270434.1 | 4 | 962 | NP_001257363.1 | |
| NM_004642.3 | 4 | 1453 | NP_004633.1 | |
| NR_073007.1 | 4 | 1036 | ||
| NR_073008.1 | 4 | 1092 | ||
| AA477326.1 | 1 | 203 | ||
| AB006077.1 | 4 | 1009 | BAA22937.1 | |
| AF006484.1 | 4 | 1453 | AAC77831.1 | |
| AI139790.1 | 1 | 175 | ||
| BC034717.1 | 4 | 952 | AAH34717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826532 | Chr.12:123259630 - 123261596 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map