Assay Details
Target Gene Details
Entrez Gene ID: | 10786 |
Gene Name: | solute carrier family 17 member 3 |
Gene Aliases: |
GOUT4, NPT4, UAQTL4 |
Location: |
Chr.6:25845100-25874243 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A3 | NM_001098486.1 | NP_001091956.1 | ||
| NM_006632.3 | NP_006623.2 | |||
| AK296070.1 | 5 | 1004 | ||
| AK298271.1 | ||||
| AK298297.1 | 5 | 950 | ||
| AK298367.1 | 7 | 905 | ||
| BC017952.1 | AAH17952.1 | |||
| CB163247.1 | ||||
| U90545.1 | AAB53423.1 |
More Information
Set Membership: |
|

Genomic Map